Learn More
LUNA NANOTECH INC Rnase A, Dnase Free, 20 Mg/ML, 48 ML
Supplier:  LUNA NANOTECH INC GRA048
Encompass_Preferred
RNase A, DNase Free, 20 mg/mL (>70U/mg), 48 mL
RNase A degrades single-stranded RNA at the C and U residues by cleaving the phosphodiester bond between the 5’-ribose of the nucleotide and the phosphate group that is connected to the 3’-ribose of the neighboring pyrimidine.
RNase A (Ribonuclease A) is commonly used in plasmid DNA and genomic DNA purification to remove contaminating RNA from DNA preparation. RNase A is also used in RNase protection assays and RNA sequence analysis.
RNase A is isolated from bovine pancreas and heat treated to inactivate any potential DNAse contamination.
Enzyme activity: >70U/mg
Catalog No. 50-256-6985