Learn More
Bioworld RNASE-ARREST BUFFER
Supplier:  Bioworld 107600441
Encompass_Preferred
RNase-ARREST BUFFER RNase Arrest Buffer is an optimized combination of various chaotropic agents and RNase inhibitors, which inhibits RNase in 5-10 seconds. Arrest Extraction Buffer may be used in conjunction with any RNA extraction meth...
Catalog No. 50-196-895
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.