Learn More
New England Biolabs, Inc. RNase I – 25000 units
SDP
Supplier:  New England Biolabs, Inc. M0243L
Encompass
Ribonuclease If (RNase If) is an RNA endonuclease which will cleave at all RNA dinucleotide bonds leaving a 5'hydroxyl and 2',3' cyclic monophosphate. It has a preference for single-stranded RNA over double-stranded RNA. RNase If is a recombinant protein fusion of RNase I (from E. coli) and maltose-binding protein. It has identical activity to RNase I.
Catalog No. 50-811-658