Learn More
Meridian Life Science RNase Inhibitor Glycerol Free, 50ML
SDP
Supplier:  Meridian Life Science MDX12050
Encompass_Preferred
RNase Inhibitor (Glycerol-Free) is from a recombinant protein that inhibits different RNases (A, B, C) by binding non-covalently in a 1:1 ratio. With an association constant of 10^14 M, it is useful in any applications where the presence of RNases is a potential problem. RNase Inhibitor (Glycerol-Free) is tested for activity, SDS-PAGE purity and the absence of endonucleases, nickases and exonuclease. RNase Inhibitor (Glycerol Free) delivers excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization for ambient temperature storage and shipment.
Catalog No. 50-008-56297