Learn More
Apexbio Technology LLC RNase Inhibitor, Murine, 3000U. Cas: MFCD: NA. RNase Inhibitor, Murine specifically inhibits RNases A, B and C to protect RNA from degradation.
SDP
Supplier:  Apexbio Technology LLC K10463000
Encompass_Preferred
RNase Inhibitor, Murine is a 50 kD recombinant protein isolated from E. coli containing the ribonuclease inhibitor gene originally from mouse. Our product RNase inhibitor has specificity for RNases A, B and C, but couldnt inhibit RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. It can bind several kinds of RNase non-covalently in a 1:1 ratio with high affinity. Especially, according to previous researches, no inhibition for enzyme activity is observed when RNase Inhibitor is used within AMV or M-MuLV reverse transcriptases, taq DNA polymerase or SP6/ T7/ T3 RNA polymerases, means it can be taken in various experiments to prevent RNA degradation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8276