Learn More
LGC GENOMICS LLC (Lucigen) Ribonuclease R (RNase R), 250 U @ 20 U/LA 3 to 5 exoribonuclease that digests all linear RNAs except lariat or circular RNA structures. Manufactured under ISO-13485.
Supplier:  LGC GENOMICS LLC RNR07250
Encompass
Ribonuclease R (RNase R) from E. coli is a magnesium-dependent 35 exoribonuclease that digests essentially all linear RNAs but does not digest lariat or circular RNA structures.Specific: Digests all linear RNAs but does not digest lariat or circular RNA structures, or double stranded RNA with 3' overhangs shorter than seven nucleotidesHeat Inactivated: Heat at 65 ¿C for 20 minutes to kill enzyme activityValuable: Use the unique properties of this exoribonuclease to study alternative splicing and gene expressionRibonuclease R includes both the enzyme and an optimised 10X Reaction Buffer.
Catalog No. NC1163864