Learn More
ABclonal Technology RNASEH2C Rabbit pAb
SDP
Supplier:  ABclonal Technology A13884500UL
Encompass_Preferred
This gene encodes a ribonuclease H subunit that can cleave ribonucleotides from RNADNA duplexes. Mutations in this gene cause Aicardi-Goutieres syndrome-3, a disease that causes severe neurologic dysfunction. A pseudogene for this gene has been identified on chromosome Y, near the sex determining region Y (SRY) gene.
Catalog No. 50-001-16208