Learn More
ABclonal Technology RNF125 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15165500UL
Encompass_Preferred
This gene encodes a novel E3 ubiquitin ligase that contains a RING finger domain in the N-terminus and three zinc-binding and one ubiquitin-interacting motif in the C-terminus. As a result of myristoylation, this protein associates with membranes and is primarily localized to intracellular membrane systems. The encoded protein may function as a positive regulator in the T-cell receptor signaling pathway.
Catalog No. 50-001-04783