Learn More
Aviva Systems Biology Corp RNF130 Antibody - N-terminal region (ARP32914_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32914_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RNF130. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RNF130. Alternate names: GP, G1RZFP, GOLIATHApplicationWB. Species reactivity: Cow, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54818