Learn More
ABclonal Technology RNF181 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14405500UL
Encompass_Preferred
RNF181 binds the integrin alpha-IIb (ITGA2B, MIM 607759)/beta-3 (ITGB3, MIM 173470) complex and has E3 ubiquitin ligase activity (Brophy et al., 2008 [PubMed 18331836]).
Catalog No. 50-001-06998