Learn More
Medchemexpress LLC Ro 10-5824 dihydrochloride | 189744-94-3 | 99.9% | 353.29 g/mol | C17H22Cl2N4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101384A100MG
Encompass_Preferred
Ro 10-5824 dihydrochloride is a selective partial agonist at the dopamine D4 receptor (Ki = 5.2 nM). Supplied as the dihydrochloride salt in solid form for research use, it is used in receptor pharmacology, GPCR functional assays, and screening applications requiring selective D4 modulation. Chemical identity: C17H22Cl2N4 (CAS 189744-94-3).
  • Selective partial agonist at dopamine D4 receptor (Ki = 5.2 nM).
  • High reported purity (99.86%).
  • Solid form for convenient handling and formulation.
  • Stable when stored sealed and away from moisture; follow recommended temperature storage.
  • Suitable for receptor pharmacology, GPCR assays, and screening workflows.
  • Characterized by CAS number and molecular weight for traceability.
Catalog No. 50-002-12830