Learn More
Medchemexpress LLC HY-100927 5mg , Ro 20-1724 CAS:29925-17-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009275MG
Encompass_Preferred
HY-100927 5mg Ro 20-1724 CAS: 29925-17-5 Ro 20-1724 (Ro 20-174) is a potent inhibitor of cAMP-specific phosphodiesterase (PDE4/PDE IV) with a Ki of 1930 nM. Neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1611