Learn More
Cayman Chemical Ro 25-6981maleate 10mg
SDP
Supplier:  Cayman Chemical 1457810 MG
Encompass_Preferred
A potent, selective activity-dependent blocker of NMDA receptors containing the NR2B subunit (IC50 = 9 nM); displays neuroprotectant effects in cortical neurons in vitro (IC50 = 0.04-0.4 μM)
Catalog No. 50-297-1116