Learn More
TARGETMOL CHEMICALS INC RO 31-8220 MESYLATE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T664310MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ro 31-8220 Mesylate (Bisindolylmaleimide IX mesylate) is a pan-PKC inhibitor for PKC-(alpha)/(beta)I/(beta)II/(gamma)/(epsilon) (IC50 5/24/14/27/24 nM) and also shows potent inhibition against MSK1 MAPKAP-K1b S6K1 and GSK3(beta). purity: 99%
Catalog No. 50-257-9321