Learn More
Abcam Ro 31-8220 mesylate (Bisindolylmaleimide I x ), Protein kinase inhibitor, 10MG
Supplier:  Abcam AB12037410MG
Encompass_Preferred
MW 553.7 Da, Purity >98%. Widely used as a cell-permeable, reversible, competitive protein kinase C inhibitor. Also potently inhibits wide range of kinases inlcuding RSK1, RSK2, PRK2, PKCα, PKD1, MSK1, GSK3β, CDK2-cyclin A and PIM3, as well as PKCα. Also available in simple stock solutions (ab146723) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7640