Learn More
Enzo Life Sciences Bisindolylmaleimide X. hydrochloride (5mg). CAS: 145317-11-9
SDP
Supplier:  Enzo Life Sciences ALX270057M005
Encompass_Preferred
Selective inhibitor of protein kinase C (PKC). Alternative name: Ro 31-8425. Formula: C26H24N4O2 . HCl. MW: 424.5 . 36.5. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8393