Learn More
Medchemexpress LLC Ro 41-1049 hydrochloride | 127917-66-2 | MFCD00900596 | 100.0% | 301.77 | C12H13ClFN3OS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100027A100MG
Encompass_Preferred
Ro 41-1049 hydrochloride is a reversible and selective monoamine oxidase A (MAO-A) inhibitor used as a research reagent in neuropharmacology and biochemical studies. It is supplied as a white to off-white solid of high purity and is appropriate for both in vitro and in vivo experimental work.
  • Selective reversible inhibition of MAO-A for mechanism-of-action studies.
  • High purity suitable for analytical and pharmacological assays.
  • Solid form simplifies handling and storage.
  • Available in small research-scale quantities for dose-ranging studies.
  • Well-characterized compound with CAS 127917-66-2 and a defined molecular weight.
  • Useful for neurochemical and behavioral pharmacology experiments.
Catalog No. 50-002-15668