Learn More
Selleck Chemical LLC RO 5028442 S0007-25MG
SDP
Supplier:  Selleck Chemical LLC S000725MG
Encompass_Preferred
RO 5028442 (RG7713) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM for hV1a and 39 nM for mV1a
Catalog No. 50-000-40074