Learn More
Selleck Chemical LLC Ro 61-8048
SDP
Supplier:  Selleck Chemical LLC S817210MM/1ML
Encompass_Preferred
Ro 61-8048 is a high-affinity kynurenine 3-hydroxylase (KMO) inhibitor with IC50 and Ki of 37 nM and 4 8 nM respectively
Catalog No. 50-000-01704