Learn More
Medchemexpress LLC Ro 61-8048 | 199666-03-0 | 99.8% | 421.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY12347
Encompass_Preferred
Ro 61-8048 is an orally active and selective inhibitor of kynurenine 3-hydroxylase, demonstrating an IC50 of 37 nM. This compound significantly increases extracellular kynurenic acid concentrations and has shown potential in reducing QUIN neo-formation and the severity of dystonia in experimental models.
  • Orally active and selective kynurenine 3-hydroxylase inhibitor
  • Increases extracellular kynurenic acid concentrations
  • Reduces QUIN neo-formation
  • Inhibits cerebral enzyme activity in vivo
  • Reduces severity of dystonia in dtsz hamsters
  • Appearance is a solid, off-white to light yellow in color
Catalog No. 50-003-64794