Learn More
Medchemexpress LLC Ro 90-7501 | 293762-45-5 | 99.1% | 340.38 | C20H16N6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1032415MG
Encompass_Preferred
Ro 90-7501 is a small-molecule research compound (C20H16N6, MW 340.38, CAS 293762-45-5) that inhibits amyloid β42 fibril assembly and reduces Aβ42-induced cytotoxicity. It also inhibits ATM phosphorylation and DNA repair, modulates innate antiviral responses, inhibits protein phosphatase 5, and has reported radiosensitizing activity in cellular models.
  • Inhibits amyloid β42 fibril assembly (reported EC50 ≈ 2 μM).
  • Inhibits ATM phosphorylation and impairs DNA repair.
  • Inhibits protein phosphatase 5 (PP5) in a TPR-dependent manner.
  • Shows radiosensitizing activity in certain cancer cell models.
  • High reported purity (99.1%).
  • Supplied as a 5 mg research sample for laboratory studies.
Catalog No. 50-002-46992