Learn More
Medchemexpress LLC Ro 90-7501 | 293762-45-5 | 99.1% | 340.38 g/mol | C20H16N6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10324110MM 1ML
Encompass_Preferred
Ro 90-7501 is a small-molecule inhibitor of amyloid β42 fibril assembly supplied for research use. It is available as a solid and as a 10 mM solution in DMSO, with reported purity of about 99.1% and molecular weight 340.38 g/mol.
  • inhibits amyloid β42 fibril assembly
  • available as 10 mM solution in DMSO and multiple solid sizes
  • high purity suitable for biological assays
  • stable under recommended cold storage conditions
  • commonly used in studies of Aβ aggregation and related cellular assays
Catalog No. 50-002-47204