Learn More
Medchemexpress LLC Ro 90-7501 | 293762-45-5 | MFCD00367977 | 99.1% | 340.38 g·mol⁻1 | C20H16N6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10324150MG
Encompass_Preferred
Ro 90-7501 is a small-molecule research compound that inhibits amyloid β42 (Aβ42) fibril assembly and reduces Aβ42-induced cytotoxicity. It has reported activities including inhibition of ATM phosphorylation and protein phosphatase 5 (PP5), enhancement of antiviral signaling, and radiosensitizing effects in cancer cells. Purity is 99.1% and molecular formula is C20H16N6.
  • Inhibits amyloid β42 fibril assembly (EC50 ≈ 2 μM).
  • Reduces Aβ42-induced cytotoxicity in cell-based assays.
  • Inhibits ATM phosphorylation and PP5 activity.
  • Enhances TLR3/RLR-induced IFN-β expression and antiviral responses.
  • Available in multiple pack sizes, including 50 MG.
Catalog No. 50-002-46034