Learn More
Medchemexpress LLC HY-11102 50mg , RO4929097 CAS:847925-91-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1110250MG
Encompass_Preferred
HY-11102 50mg RO4929097 CAS: 847925-91-1 RO4929097 (RG-4733) is a γ secretase inhibitor with IC50 of 4 nM, inhibiting cellular processing of Aβ40 and Notch with EC50 of 14 nM and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9454