Learn More
Selleck Chemical LLC RO4929097 50mg 847925-91-1
SDP
Supplier:  Selleck Chemical LLC S157550MG
Encompass_Preferred
RO4929097 is a ? secretase inhibitor with IC50 of 4 nM in a cell-free assay, inhibiting cellular processing of A?40 and Notch with EC50 of 14 nM and 5 nM, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-754-3