Learn More
Medchemexpress LLC Ro4987655 | 874101-00-5 | 99.0% | 565.28 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14719
Encompass_Preferred
Ro4987655 is an orally active and highly selective MEK inhibitor with an IC50 of 5.2 nM for inhibition of MEK1/MEK2. It potently inhibits the mitogen-activated protein kinase signaling pathway activation and tumor cell growth, and inhibits proliferation of NCI-H2122 cells in a dose-dependent manner. This compound appears as a solid, white to off-white in color.
  • Highly selective MEK1/MEK2 inhibitor
  • Potently inhibits MAPK signaling pathway and tumor cell growth
  • Inhibits proliferation of NCI-H2122 cells
  • Achieves complete tumor regression in xenograft models with single-agent oral administration
  • Rapidly absorbed with dose-proportional exposures
Catalog No. 50-003-63123