Learn More
Medchemexpress LLC RO5-3335 25G

Supplier: Medchemexpress LLC HY10847025G
- CAS Number: 30195-30-3
- Molecular Formula: C13 H10 Cl N3 O
- Molecular Weight: 259.69 g/mol
- Purity: 99.35%
- Appearance: Solid
- Color: Off-white to light yellow
- Target: RUNX1-CBFβ interaction
- Mechanism of Action: Inhibitor of RUNX1-CBFβ interaction
- Activity: CBF inhibitor, Tat antagonist
- IC50 (ME-1 cells): 1.1 µM
- Solubility: 100 mM in DMSO
- Storage (Powder): -20°C for 3 years
- Storage (Stock Solution): -80°C for 2 years
- Application: Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.