Learn More
Medchemexpress LLC HY-16902 10mg , RO9021 CAS:1446790-62-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1690210MG
Encompass_Preferred
HY-16902 10mg RO9021 CAS: 1446790-62-0 RO9021 is an orally bioavailable, novel ATP-competitive inhibitor of SYK, with an average IC50 of 5.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1496