Learn More
TARGETMOL CHEMICALS INC RO9021 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167765MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. RO9021 is an orally bioavailable novel ATP-competitive SYK inhibitor (average IC50 5.6 nM). purity: 97%
Catalog No. 50-258-1154