Learn More
Medchemexpress LLC Robenidine-d8 hydrochloride | 1173097-77-2 | 99.0% | 378.71 g/mol | C15H6D8Cl3N5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB2157S10MG
Encompass_Preferred
Robenidine-d8 hydrochloride is the deuterium-labeled hydrochloride salt of robenidine, provided as a high-purity analytical standard for laboratory research. It is used in biochemical and microbiological studies, including investigations of antimicrobial activity and analytical method development.
  • Deuterium-labeled analog of robenidine.
  • Hydrochloride salt form for improved stability.
  • High purity suitable for analytical applications.
  • Provided in a small research quantity for laboratory use.
  • Useful in antimicrobial and method-development studies.
Catalog No. 50-002-97348