Learn More
Apexbio Technology LLC Robenidine hydrochloride 25875-50-7 100mg
SDP
Supplier:  Apexbio Technology LLC C6934100
Encompass_Preferred
Robenidine hydrochloride (CAS 25875-50-7) is a small-molecule antimicrobial agent with activity against methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant Enterococcus (VRE) It is designed to inhibit these bacterial pathogens thereby interfering with microbial survival Robenidine hydrochloride exerts its biological activity primarily through disruption of microbial cell membrane integrity In in vitro studies Robenidine hydrochloride demonstrates inhibitory activity with MIC50 values of 8 1 M for MRSA and 4 7 M for VRE Based on these pharmacological properties Robenidine hydrochloride holds research potential in the fields of microbial infection drug resistance and the development of novel anti-infective strategies
Catalog No. 50-000-90786