Learn More
Selleck Chemical LLC Robenidine Hydrochloride S4876-25mg
SDP
Supplier:  Selleck Chemical LLC S487625MG
Encompass_Preferred
Robenidine Hydrochloride (Dexium Nugatex Rasexinium Toraxine) is an anticoccidial agent mainly for poultry
Catalog No. 50-311-0837