Learn More
Selleck Chemical LLC Roblitinib (FGF401) 5mg 1708971-55-4
SDP
Supplier:  Selleck Chemical LLC S85485MG
Encompass_Preferred
Roblitinib (FGF401) is an FGFR4-selective inhibitor with an IC50 of 1.1 nM. It binds in a reversible covalent manner to the FGFR4 kinase domain and shows at least 1,000 fold selectivity against of panel of 65 kinases in biochemical assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8205