Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit, For processing: 30 mL buffy coat processed
SDP
Supplier:  STEMCELL Technologies 17878RF
Encompass
The EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit is designed to isolate CD14+ cells from freshly prepared buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD14 antigen is strongly expressed on monocytes and is weakly expressed on granulocytes. It is also expressed on most tissue macrophages.This product replaces the EasySep™ Human Buffy Coat CD14 Positive Selection Kit (Catalog #18088) for even faster cell isolations.
Catalog No. 50-197-4583