Learn More
STEMCELL Technologies RoboSep™ Human Naïve B Cell Enrichment Kit with Filter Tips, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19254RF
Encompass
The EasySep™ Human Naïve B Cell Enrichment Kit is designed to isolate naïve B cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-naïve B cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.
Catalog No. 50-197-4682