Learn More
STEMCELL Technologies RoboSep™ Mouse PE Positive Selection Kit II, For processing: 5 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 17696RF
Encompass
The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This kit replaces the EasySep™ Mouse PE Positive Selection Kit (Catalog #18554).For highly purified particle-free cells, we recommend the EasySep™ Release Mouse PE Positive Selection Kit (Catalog #17656). (https://www.stemcell.com/products/easysep-release-mouse-pe-positive-selection-kit.html)
Catalog No. 50-197-4556