Learn More
Medchemexpress LLC HY-19356 1mg Medchemexpress, Rocaglamide CAS:84573-16-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY193561MG
Encompass_Preferred
Medchemexpress, HY-19356 1mg Rocaglamide CAS:84573-16-0 Rocaglamide (Roc-A) is isolated from the genus Aglaia and can be used for coughs, injuries, asthma and inflammatory skin diseases. Rocaglamide is a potent inhibitor of NF-κB activation in T-cells. Rocaglamide is a potent and selective heat shock factor 1 (HSF1) activation inhibitor with an IC50 of ~50 nM. Rocaglamide inhibits the function of the translation initiation factor eIF4A. Rocaglamide also has anticancer properties in leukemia[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9278