Learn More
Rochelle Salt Stabilizer APHA for Ammonia Nitrogen, Reagents
Rochelle Salt Stabilizer APHA for Ammonia Nitrogen, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$148.00
Specifications
| Chemical Name or Material | Rochelle Salt Stabilizer APHA for Ammonia Nitrogen |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 6381-59-5 |
| Boiling Point | 101°C |
| Grade | Reagent |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-074
|
Reagents Holdings Llc
CR0050001A |
1 L |
Each of 1 for $148.00
|
|
|||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.