Learn More
Rochelle Salt Stabilizer APHA for Ammonia Nitrogen, Reagents
Specifications
Specifications
| Chemical Name or Material | Rochelle Salt Stabilizer APHA for Ammonia Nitrogen |
| Color | Colorless |
| CAS | 7732-18-5, 6381-59-5 |
| Boiling Point | 101°C |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.097 |
| pH | 6.5 to 8.5 |
| Quantity | 1 L |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.