Learn More
Selleck Chemical LLC Rociletinib (CO-1686) 10mg 1374640-70-6 AVL-301
SDP
Supplier:  Selleck Chemical LLC S728410MG
Encompass_Preferred
Rociletinib (CO-1686, AVL-301) is an irreversible, mutant-selective EGFR inhibitor with Ki of 21.5 nM and 303.3 nM for EGFRL858R/T790M and EGFRWT in cell-free assays, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6340