Learn More
Sino Biological Recombinant Canine RIPK1/RIP1 Protein (GST Tag), Active
Supplier:  Sino Biological R07D112G50
Encompass_Preferred
Recombinant dog RIPK1 (1-373) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3323