Learn More
Medchemexpress LLC Rock2-in-2 | 1995065-79-6 | 98.1% | 360.39 g/mol | C18H12N6OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10362050MG
Encompass_Preferred
ROCK2-IN-2 is a selective inhibitor of Rho-associated protein kinase 2 (ROCK2) described in patent US20180093978A1 (Compound A-30). It is provided for research use and is reported to have an IC50 of approximately 1 μM against ROCK2.
  • Selective inhibition of ROCK2 for target validation and signaling studies.
  • Reported IC50 of about 1 μM against ROCK2.
  • High supplier-reported purity (~98%).
  • Soluble in DMSO for in vitro assay preparation.
  • Available in small-scale quantities suitable for laboratory research.
Catalog No. 50-002-44570