Learn More
Medchemexpress LLC Rock2-in-2 | 1995065-79-6 | MFCD31692339 | 98.1% | 360.39 g·mol⁻¹ | C18H12N6OS | 25 MG

Supplier: Medchemexpress LLC HY10362025MG
ROCK2-IN-2 is a selective ROCK2 inhibitor (IC50 <1 μM) described in patent US20180093978A1 (Compound A-30). It is supplied as a research reagent for in vitro and cellular studies, with reported purity 98.05% and molecular weight 360.39 g·mol⁻¹. The compound is provided as a solid and is soluble in DMSO.
- Selective ROCK2 inhibition with IC50 <1 μM.
- Reported purity 98.05%.
- Chemical formula C18H12N6OS; molecular weight 360.39 g·mol⁻¹.
- Soluble in DMSO.
- Available in small-scale packs, including 25 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.