Learn More
Strem, An Ascensus Company CAS# 1262046-34-3. 500mg. 2-(Di-t-butylphosphino)-3-methoxy-6-methyl-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% RockPhos. MFCD20922895
SDP
Supplier:  Strem, An Ascensus Company 151168500MG
Encompass
CAS# 1262046-34-3. 500mg. 2-(Di-t-butylphosphino)-3-methoxy-6-methyl-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% RockPhos. MFCD20922895. Molecular Weight: 468.69. Molecular Formula: C31H49OP. Color/form: white xtl. Strem# 15-1168. http://www.strem.com/catalog/v/15-1168/
Catalog No. 50-201-6651