Learn More
Cayman Chemical RocuronIumbromIde 10mg
SDP
Supplier:  Cayman Chemical 2369810 MG
Encompass_Preferred
A non-depolarizing muscle relaxant; selectively induces neuromuscular blockade in cats (ED50s = 153, 185, 1,329, and 4,119 μg/kg for tibialis, soleus, and vagus muscles, and nictitating membranes, respectively; has a more rapid onset time than vecuronium
Catalog No. 50-299-4664