Learn More
Grainger Rod Stock Brass 3/32 in x 3 ft PK5
Supplier:  Grainger 16NH92
Encompass_Preferred
This 3/32 inch diameter brass rod stock from Grainger is 3 feet long and comes in a pack of 5 rods. It is made from C36000 free-cutting brass alloy which contains lead and is machinable. The rods have a smooth surface finish and meet ASTM B16 standards. They can be cut, machined, or otherwise fabricated for various applications where a corrosion resistant, strong copper alloy is needed. The brass alloy has good formability.
Catalog No. 50-276-4526