Learn More
TARGETMOL CHEMICALS INC ROFLEPONIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T343835MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Rofleponide is a synthetic glucocorticosteroid with a high affinity for the rat thymus glucocorticoid receptor. purity: 99%
Catalog No. 50-258-0264