Learn More
Cayman Chemical RoflumIlast-d4 1mg
SDP
Supplier:  Cayman Chemical 182601 MG
Encompass_Preferred
An internal standard for the quantification of roflumilast by GC- or LC-MS
Catalog No. 50-296-8371