Learn More
Cayman Chemical RoflumIlast-d4 500ug
SDP
Supplier:  Cayman Chemical 18260500 UG
Encompass_Preferred
An internal standard for the quantification of roflumilast by GC- or LC-MS
Catalog No. 50-296-6446