Learn More
Medchemexpress LLC Roflumilast-d4 N-Oxide | 1794760-31-8 | C17H10D4Cl2F2N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15455S15MG
Encompass_Preferred
Roflumilast-d4 N-Oxide is the deuterium labeled version of Roflumilast, a selective PDE4 inhibitor with IC50s of 0.7, 0.9, 0.7, and 0.2 nM for PDE4A1, PDEA4, PDEB1, and PDEB2, respectively. It does not affect PDE1, PDE2, PDE3, or PDE5 isoenzymes from various cells. This compound is intended for research use only.
  • Deuterium labeled version of roflumilast
  • Selective PDE4 inhibitor
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-76276